APO455828636 Apollo Scientific Ltd

2'-(Allyloxy)-6'-hydroxyacetophenone

  • MDL number MFCD00100491
  • Cas Number : 23226-84-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 45.5-46.5
Synonym 1-[2-(Allyloxy)-6-hydroxyphenyl]ethan-1-one
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=18229

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=18229
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart