APO455828643 Apollo Scientific Ltd

5-Aminobenzene-1,3-diol hydrochloride

  • MDL number MFCD00662897
  • Cas Number : 6318-56-5
  • Molecular Weight : 161.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 300
Synonym 3,5-Dihydroxyaniline hydrochloride, 5-Aminoresorcinol hydrochloride
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-aminobenzene-13-diol-hydrochloride

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-aminobenzene-13-diol-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart