APO455828644 Apollo Scientific Ltd

Ethyl 6-iodochromone-2-carboxylate

  • MDL number MFCD00100378
  • Cas Number : 35204-44-5
  • Molecular Weight : 344.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 148-150
Iupac Name ethyl 6-iodo-4-oxo-4H-chromene-2-carboxylate
Inchi Code 1S/C12H9IO4/c1-2-16-12(15)11-6-9(14)8-5-7(13)3-4-10(8)17-11/h3-6H,2H2,1H3
Inchi Key HUJFMOAXSPSGDU-UHFFFAOYSA-N

Description

Synonym Ethyl 6-iodo-4-oxo-4H-chromene-2-carboxylate, Ethyl 6-iodo-4-oxo-4H-1-benzopyran-2-carboxylate
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 148-150
Product Link https://store.apolloscientific.co.uk/product/ethyl-6-iodochromone-2-carboxylate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-6-iodochromone-2-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart