APO455828645 Apollo Scientific Ltd

4-(But-1-yl)biphenyl

  • MDL number MFCD00100665
  • Cas Number : 37909-95-8
  • Molecular Weight : 210.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp >110
bp 44414
Iupac Name 4-butyl-1,1'-biphenyl
density 0.953
Inchi Code 1S/C16H18/c1-2-3-7-14-10-12-16(13-11-14)15-8-5-4-6-9-15/h4-6,8-13H,2-3,7H2,1H3
Inchi Key JZDZRFNMDCBTNS-UHFFFAOYSA-N

Description

Melting Point >110
Synonym 1-(Biphenyl-4-yl)butane
Purity 98%
Density 0.953
Flash Point 318-319
Storage temp. Ambient Temperature
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 44414
Product Link https://store.apolloscientific.co.uk/product/4-but-1-ylbiphenyl

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-but-1-ylbiphenyl
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart