APO455828653 Apollo Scientific Ltd

1-(4-Hydroxyphenyl)butan-1-one

  • MDL number MFCD00020173
  • Cas Number : 1009-11-6
  • Molecular Weight : 164.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 89-93
Iupac Name 1-(4-hydroxyphenyl)-1-butanone
Inchi Code 1S/C10H12O2/c1-2-3-10(12)8-4-6-9(11)7-5-8/h4-7,11H,2-3H2,1H3
Inchi Key GFBLPULLSAPXDC-UHFFFAOYSA-N

Description

Flash Point 148-150/11.5mm
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 89-93
Product Link https://store.apolloscientific.co.uk/product/1-4-hydroxyphenylbutan-1-one

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/1-4-hydroxyphenylbutan-1-one
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart