APO455828655 Apollo Scientific Ltd

3-Hydroxy-4-iodobenzaldehyde

  • MDL number MFCD00114697
  • Cas Number : 135242-71-6
  • Molecular Weight : 248.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 126-128
Iupac Name 3-hydroxy-4-iodobenzaldehyde
Inchi Code 1S/C7H5IO2/c8-6-2-1-5(4-9)3-7(6)10/h1-4,10H
Inchi Key IHLOHISMHMTTAA-UHFFFAOYSA-N

Description

Synonym 5-Formyl-2-iodophenol
Purity 98%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 126-128
Product Link https://store.apolloscientific.co.uk/product/3-hydroxy-4-iodobenzaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-hydroxy-4-iodobenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart