APO455828658 Apollo Scientific Ltd

2,6-Dibromonitrobenzene

  • MDL number MFCD09998726
  • Cas Number : 13402-32-9
  • Molecular Weight : 280.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78-82
Synonym 1,3-Dibromo-2-nitrobenzene
Purity 95
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/26-dibromonitrobenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/26-dibromonitrobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart