APO455828660 Apollo Scientific Ltd

3-Bromo-2-nitroaniline

  • MDL number MFCD08063248
  • Cas Number : 7138-15-0
  • Molecular Weight : 217.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromo-2-nitroaniline
Inchi Code 1S/C6H5BrN2O2/c7-4-2-1-3-5(8)6(4)9(10)11/h1-3H,8H2
Inchi Key GLKHLBAXHLAQBJ-UHFFFAOYSA-N

Description

Synonym 2-Amino-6-bromonitrobenzene
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-2-nitroaniline

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-2-nitroaniline
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart