APO455828671 Apollo Scientific Ltd

2-Amino-6-ethylpyridine

  • MDL number MFCD00218265
  • Cas Number : 21717-29-3
  • Molecular Weight : 122.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-ethyl-2-pyridinylamine
Inchi Code 1S/C7H10N2/c1-2-6-4-3-5-7(8)9-6/h3-5H,2H2,1H3,(H2,8,9)
Inchi Key JXKAUUVMXZIJNZ-UHFFFAOYSA-N

Description

Synonym 6-Ethylpyridin-2-amine
Purity 95%
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-6-ethylpyridine

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-6-ethylpyridine
pictograms GHS06
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart