APO455828677 Apollo Scientific Ltd

3-Methyl-N-[(1E)-3-(4-methylphenyl)-3-oxoprop-1-en-1-yl]pyridinium chloride

  • MDL number MFCD00204658
  • Cas Number : 19518-27-5
  • Molecular Weight : 273.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N-[(1E)-3-(4-Methylphenyl)-3-oxoprop-1-en-1-yl]-3-picolinium chloride
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-methyl-n-1e-3-4-methylphenyl-3-oxoprop-1-en-1-ylpyridinium-chloride

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-methyl-n-1e-3-4-methylphenyl-3-oxoprop-1-en-1-ylpyridinium-chloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart