APO455828682 Apollo Scientific Ltd

Hex-1-yl sulphanylacetate

  • MDL number MFCD00022089
  • Cas Number : 20292-01-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Hex-1-yl mercaptoacetate, Hex-1-yl 2-sulphanylethanoate
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=18487

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=18487
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart