APO455828688 Apollo Scientific Ltd

4,5-Dibromothiophene-2-carboxylic acid

  • MDL number MFCD00039682
  • Cas Number : 6324-10-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Carboxy-2,3-dibromothiophene, 4,5-Dibromo-2-thenoic acid
Storage temp. Store at 2-8
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=18629

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=18629
Availability details for
Quantity
  • Availability
  • Add to Cart