APO455828693 Apollo Scientific Ltd

3-[(Methylamino)methyl]azetidine hydrochloride, 3-BOC protected

  • MDL number MFCD09998721
  • Cas Number : 1170019-97-2
  • Molecular Weight : 236.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 3-azetidinylmethyl(methyl)carbamate hydrochloride
Inchi Code 1S/C10H20N2O2.ClH/c1-10(2,3)14-9(13)12(4)7-8-5-11-6-8;/h8,11H,5-7H2,1-4H3;1H
Inchi Key NPZOOGZMCGJGTO-UHFFFAOYSA-N

Description

Synonym tert-Butyl [(azetidin-3-yl)methyl]methylcarbamate hydrochloride, 3-{[(tert-Butoxycarbonyl)(methyl)amino]methyl}azetidine hydrochloride
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-methylaminomethylazetidine-hydrochloride-3-boc-protected

Safety Information

signalword Warning
pcodes P280;P271;P260
MSDS https://store.apolloscientific.co.uk/product/3-methylaminomethylazetidine-hydrochloride-3-boc-protected
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart