APO455828703 Apollo Scientific Ltd

4-(2-Aminoethyl)piperidine, 4-BOC protected

  • MDL number MFCD01318373
  • Cas Number : 165528-81-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 48-50
Synonym tert-Butyl [2-(piperidin-4-yl)ethyl]carbamate, N-(tert-Butoxycarbonyl)-2-(piperidin-4-yl)ethylamine
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=40161

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=40161
pictograms GHS05;GHS07
hstatements H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart