APO455828706 Apollo Scientific Ltd

N-(4-Aminophenyl)-N-methylacetamide

  • MDL number MFCD00007859
  • Cas Number : 119-63-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N1-Acetyl-N1-methylbenzene-1,4-diamine, N1-Acetyl-N1-methylphenylene-1,4-diamine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/n-4-aminophenyl-n-methylacetamide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/n-4-aminophenyl-n-methylacetamide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart