APO455828707 Apollo Scientific Ltd

2,6-Dibromoisonicotinic acid

  • MDL number MFCD09743987
  • Cas Number : 2016-99-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,6-Dibromopyridine-4-carboxylic acid
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/26-dibromoisonicotinic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/26-dibromoisonicotinic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart