APO455828719 Apollo Scientific Ltd

4-Cyclohexylbenzenesulphonyl chloride

  • MDL number MFCD00625713
  • Cas Number : 56354-57-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 42-52
Synonym [4-(Chlorosulphonyl)phenyl]cyclohexane
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-cyclohexylbenzenesulphonyl-chloride

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-cyclohexylbenzenesulphonyl-chloride
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart