APO455828720 Apollo Scientific Ltd

4-Amino-N,N-dimethylbenzenesulphonamide

  • MDL number MFCD00031428
  • Cas Number : 1709-59-7
  • Molecular Weight : 200.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 168-172
Iupac Name 4-amino-N,N-dimethylbenzenesulfonamide
Inchi Code 1S/C8H12N2O2S/c1-10(2)13(11,12)8-5-3-7(9)4-6-8/h3-6H,9H2,1-2H3
Inchi Key BABGMPQXLCJMSK-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 168-172
Product Link https://store.apolloscientific.co.uk/product/4-amino-nn-dimethylbenzenesulphonamide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-amino-nn-dimethylbenzenesulphonamide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart