APO455828721 Apollo Scientific Ltd

1-(5-Bromo-1H-indol-3-yl)-2-[4-(pyrimidin-2-yl)piperazin-1-yl]ethane-1,2-dione

  • MDL number MFCD00829501
  • Cas Number : 256417-42-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Bromo-3-{oxo[4-(pyrimidin-2-yl)piperazin-1-yl]acetyl}-1H-indole, 1-[(5-Bromo-1H-indol-3-yl)(oxo)acetyl]-4-(pyrimidin-2-yl)piperazine
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=18992

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=18992
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart