APO455828727 Apollo Scientific Ltd

3-(Methoxycarbonyl)-5-[methyl(methylsulphonyl)amino]benzoic acid

  • MDL number MFCD09954475
  • Cas Number : 695215-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Methyl 5-[methyl(methylsulphonyl)amino]isophthalate, N-[3-Carboxy-5-(methoxycarbonyl)phenyl]-N-methylmethanesulphonamide
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=42330

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=42330
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart