APO455828733 Apollo Scientific Ltd

2-Aminothiophene, N-BOC protected

  • MDL number MFCD02677742
  • Cas Number : 56267-50-6
  • Molecular Weight : 199.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 151-152
Synonym tert-Butyl (thien-2-yl)carbamate, 2-[(tert-Butoxycarbonyl)amino]thiophene
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-aminothiophene-n-boc-protected

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-aminothiophene-n-boc-protected
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart