APO455828740 Apollo Scientific Ltd

3,5-Dimethylisoxazole-4-carbonitrile

  • MDL number MFCD02681974
  • Cas Number : 31301-46-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 43-46
Synonym 4-Cyano-3,5-dimethylisoxazole, 4-Cyano-3,5-dimethyl-1,2-oxazole
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=19183

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=19183
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart