APO455828741 Apollo Scientific Ltd

5-Methylisoxazole-4-sulphonyl chloride

  • MDL number MFCD02681975
  • Cas Number : 321309-26-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(Chlorosulphonyl)-5-methylisoxazole, 5-Methyl-1,2-oxazole-4-sulphonyl chloride
Physical Form Liquid-Viscous
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 56-58/0.7mm
Product Link https://store.apolloscientific.co.uk/product/5-methylisoxazole-4-sulphonyl-chloride

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-methylisoxazole-4-sulphonyl-chloride
pictograms GHS05
hstatements H314;EUH014;EUH029
Availability details for
Quantity
  • Availability
  • Add to Cart