APO455828751 Apollo Scientific Ltd

Homopiperazin-5-one hydrochloride

  • MDL number MFCD09864983
  • Cas Number : 208245-76-5
  • Molecular Weight : 150.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1,4-diazepan-5-one hydrochloride
Inchi Code 1S/C5H10N2O.ClH/c8-5-1-2-6-3-4-7-5;/h6H,1-4H2,(H,7,8);1H
Inchi Key WOSZTROLFNNEIW-UHFFFAOYSA-N

Description

Synonym 1,4-Diazepan-5-one hydrochloride, 5-Oxo-1,4-diazepane hydrochloride
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/homopiperazin-5-one-hydrochloride

Safety Information

signalword Warning
pcodes P271;P280;P261
MSDS https://store.apolloscientific.co.uk/product/homopiperazin-5-one-hydrochloride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart