APO455828759 Apollo Scientific Ltd

4-Bromo-3,5-dimethyl-1H-pyrazole

  • MDL number MFCD00005242
  • Cas Number : 3398-16-1
  • Molecular Weight : 175.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 123-125
Iupac Name 4-bromo-3,5-dimethyl-1H-pyrazole
Inchi Code 1S/C5H7BrN2/c1-3-5(6)4(2)8-7-3/h1-2H3,(H,7,8)
Inchi Key RISOHYOEPYWKOB-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 123-125
Product Link https://store.apolloscientific.co.uk/product/4-bromo-35-dimethyl-1h-pyrazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-35-dimethyl-1h-pyrazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart