APO455828791 Apollo Scientific Ltd

Quinoline-6-carboxylic acid

  • MDL number MFCD00047613
  • Cas Number : 10349-57-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 293-296
Synonym 6-Carboxyquinoline, 6-Carboxy-1-azanaphthalene
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/quinoline-6-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/quinoline-6-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart