APO455828797 Apollo Scientific Ltd

5-Bromo-2,4-dimethyl-1,3-thiazole

  • MDL number MFCD03086099
  • Cas Number : 28599-52-2
  • Molecular Weight : 192.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-2,4-dimethyl-1,3-thiazole
Inchi Code 1S/C5H6BrNS/c1-3-5(6)8-4(2)7-3/h1-2H3
Inchi Key BSFCVAYFJQLZEU-UHFFFAOYSA-N

Description

Flash Point 114-120/15mm
Storage temp. Store at 2-8
Physical Form Liquid-Viscous
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-24-dimethyl-13-thiazole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-24-dimethyl-13-thiazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart