APO455828798 Apollo Scientific Ltd

1,3-Benzothiazole-5-carboxylic acid

  • MDL number MFCD06796380
  • Cas Number : 68867-17-4
  • Molecular Weight : 179.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 258-261(dec.)
Iupac Name 1,3-benzothiazole-5-carboxylic acid
Inchi Code 1S/C8H5NO2S/c10-8(11)5-1-2-7-6(3-5)9-4-12-7/h1-4H,(H,10,11)
Inchi Key RBIZQDIIVYJNRS-UHFFFAOYSA-N

Description

Synonym 5-Carboxy-1,3-benzothiazole
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 258-261(dec.)
Product Link https://store.apolloscientific.co.uk/product/13-benzothiazole-5-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/13-benzothiazole-5-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart