APO455828813 Apollo Scientific Ltd

1H-Imidazole-4-thiocarboxamide

  • MDL number MFCD00233357
  • Cas Number : 95962-95-1
  • Molecular Weight : 127.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 204-207
Synonym 1H-Imidazole-4-carbothioic acid amide, 4-Carbamothioyl-1H-imidazole
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1h-imidazole-4-thiocarboxamide

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1h-imidazole-4-thiocarboxamide
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart