APO455828822 Apollo Scientific Ltd

5-Bromo-2,1,3-benzoxadiazole

  • MDL number MFCD02682026
  • Cas Number : 51376-06-8
  • Molecular Weight : 199.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 73-74
Iupac Name 5-bromo-2,1,3-benzoxadiazole
Inchi Code 1S/C6H3BrN2O/c7-4-1-2-5-6(3-4)9-10-8-5/h1-3H
Inchi Key ZWDFFESFCIACQC-UHFFFAOYSA-N

Description

Synonym 5-Bromobenzofurazan
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 73-74
Product Link https://store.apolloscientific.co.uk/product/5-bromo-213-benzoxadiazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-213-benzoxadiazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart