APO455828826 Apollo Scientific Ltd

3-(Bromomethyl)-5-methylisoxazole

  • MDL number MFCD02677720
  • Cas Number : 130628-75-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(Bromomethyl)-5-methyl-1,2-oxazole
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 44/0.4mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=19349

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=19349
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart