APO455828836 Apollo Scientific Ltd

2-(Aminomethyl)benzo[b]thiophene

  • MDL number MFCD00965305
  • Cas Number : 6314-43-8
  • Molecular Weight : 163.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 58-59
Iupac Name 1-benzothien-2-ylmethanamine
Inchi Code 1S/C9H9NS/c10-6-8-5-7-3-1-2-4-9(7)11-8/h1-5H,6,10H2
Inchi Key WKYFWFHTABURGB-UHFFFAOYSA-N

Description

Synonym (Benzo[b]thiophen-2-yl)methylamine, (1-Benzothiophen-2-yl)methylamine
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 58-59
Product Link https://store.apolloscientific.co.uk/product/2-aminomethylbenzobthiophene

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-aminomethylbenzobthiophene
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart