APO455828844 Apollo Scientific Ltd

4-Benzylmorpholine-2-carboxylic acid hydrochloride

  • MDL number MFCD02682031
  • Cas Number : 135072-15-0
  • Molecular Weight : 257.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 244-248
Iupac Name 4-benzyl-2-morpholinecarboxylic acid hydrochloride
Inchi Code 1S/C12H15NO3.ClH/c14-12(15)11-9-13(6-7-16-11)8-10-4-2-1-3-5-10;/h1-5,11H,6-9H2,(H,14,15);1H
Inchi Key CEDXMALCJZSQHA-UHFFFAOYSA-N

Description

Synonym 4-Benzyl-2-carboxymorpholine hydrochloride
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 244-248
Product Link https://store.apolloscientific.co.uk/product/4-benzylmorpholine-2-carboxylic-acid-hydrochloride

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-benzylmorpholine-2-carboxylic-acid-hydrochloride
pictograms GHS06
hstatements H301+H311+H331
Availability details for
Quantity
  • Availability
  • Add to Cart