APO455828850 Apollo Scientific Ltd

1,5-Dimethyl-3-(hydroxymethyl)-1H-pyrazole

  • MDL number MFCD03659702
  • Cas Number : 153912-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 50-51
Synonym (1,5-Dimethyl-1H-pyrazol-3-yl)methanol
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 118/2mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=19395

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=19395
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart