APO455828856 Apollo Scientific Ltd

Thien-3-yl isocyanate

  • MDL number MFCD02681899
  • Cas Number : 76536-95-3
  • Molecular Weight : 125.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-isocyanatothiophene
Inchi Code 1S/C5H3NOS/c7-4-6-5-1-2-8-3-5/h1-3H
Inchi Key PVIZQOWWCLBJES-UHFFFAOYSA-N

Description

Synonym 3-Isocyanatothiophene
Flash Point 36-37/0.75mm
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/thien-3-yl-isocyanate

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/thien-3-yl-isocyanate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart