APO455828857 Apollo Scientific Ltd

3-(Aminomethyl)thiophene

  • MDL number MFCD01529872
  • Cas Number : 27757-86-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (Thien-3-yl)methylamine
Purity 95
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 139
Product Link https://store.apolloscientific.co.uk/product/3-aminomethylthiophene

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-aminomethylthiophene
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart