APO455828858 Apollo Scientific Ltd

3,4-Dihydro-2H-1,5-benzodioxepin-7-boronic acid

  • MDL number MFCD02681910
  • Cas Number : 279261-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150
Purity 90
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=19418

Safety Information

signalword Danger
pcodes P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=19418
pictograms GHS07;GHS05
hstatements H315;H319;H335;H318
Availability details for
Quantity
  • Availability
  • Add to Cart