APO455828861 Apollo Scientific Ltd

2-(Pyridin-3-yl)-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD00052304
  • Cas Number : 39067-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 283-284
Synonym 4-Carboxy-2-(pyridin-3-yl)-1,3-thiazole, 3-(4-Carboxy-1,3-thiazol-2-yl)pyridine
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=19426

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=19426
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart