APO455828864 Apollo Scientific Ltd

1,3,5-Trimethyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD00159648
  • Cas Number : 1125-29-7
  • Molecular Weight : 154.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 215-219
Synonym 4-Carboxy-1,3,5-trimethyl-1H-pyrazole
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/135-trimethyl-1h-pyrazole-4-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/135-trimethyl-1h-pyrazole-4-carboxylic-acid
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart