APO455828865 Apollo Scientific Ltd

1,3,5-Trimethyl-1H-pyrazole-4-carboxaldehyde

  • MDL number MFCD00159623
  • Cas Number : 2644-93-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 71-74
Synonym 4-Formyl-1,3,5-trimethyl-1H-pyrazole
Purity 97
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 132/12mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=19442

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=19442
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart