APO455828866 Apollo Scientific Ltd

4-(Aminomethyl)-1,3,5-trimethyl-1H-pyrazole

  • MDL number MFCD02055811
  • Cas Number : 352018-93-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (1,3,5-Trimethyl-1H-pyrazol-4-yl)methylamine
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=19445

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=19445
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart