APO455828868 Apollo Scientific Ltd

2-(3,5-Dimethyl-1H-pyrazol-4-yl)benzoic acid

  • MDL number MFCD02682016
  • Cas Number : 321309-43-7
  • Molecular Weight : 216.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(3,5-dimethyl-1H-pyrazol-4-yl)benzoic acid
Inchi Code 1S/C12H12N2O2/c1-7-11(8(2)14-13-7)9-5-3-4-6-10(9)12(15)16/h3-6H,1-2H3,(H,13,14)(H,15,16)
Inchi Key JNHKHFADRGJMJS-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-35-dimethyl-1h-pyrazol-4-ylbenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-35-dimethyl-1h-pyrazol-4-ylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart