APO455828869 Apollo Scientific Ltd

5-Methyl-2-phenyl-2H-1,2,3-triazole-4-carbonyl chloride

  • MDL number MFCD00052545
  • Cas Number : 36401-55-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105-107
Purity 97
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=19459

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=19459
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart