APO455828871 Apollo Scientific Ltd

4-(1,2,3-Thiadiazol-4-yl)benzoic acid

  • MDL number MFCD00085167
  • Cas Number : 187999-31-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 230
Synonym 4-(4-Carboxyphenyl)-1,2,3-thiadiazole
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=19464

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=19464
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart