APO455828874 Apollo Scientific Ltd

4-(1,2,3-Thiadiazol-4-yl)benzyl bromide

  • MDL number MFCD00052102
  • Cas Number : 163798-92-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-137
Synonym 4-[4-(Bromomethyl)phenyl]-1,2,3-thiadiazole
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=19467

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=19467
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart