APO455828881 Apollo Scientific Ltd

6-(Morpholin-4-yl)nicotinaldehyde

  • MDL number MFCD03659703
  • Cas Number : 173282-60-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88-89
Synonym 6-(Morpholin-4-yl)pyridine-3-carboxaldehyde, 4-(5-Formylpyridin-2-yl)morpholine
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/6-morpholin-4-ylnicotinaldehyde

Safety Information

signalword Warning
pcodes P260;P280;P301+P312
MSDS https://store.apolloscientific.co.uk/product/6-morpholin-4-ylnicotinaldehyde
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart