APO455828885 Apollo Scientific Ltd

2-Phenyl-1,3-thiazole-4-carbonyl chloride

  • MDL number MFCD02681968
  • Cas Number : 36094-04-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=19488

Safety Information

signalword Danger
pcodes P260;P280;P310
MSDS http://www.apolloscientific.co.uk/display_item.php?id=19488
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart