APO455828886 Apollo Scientific Ltd

2-Phenyl-1,3-thiazole-4-carboxaldehyde

  • MDL number MFCD02681934
  • Cas Number : 20949-81-9
  • Molecular Weight : 189.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 51-52
Iupac Name 2-phenyl-1,3-thiazole-4-carbaldehyde
Inchi Code 1S/C10H7NOS/c12-6-9-7-13-10(11-9)8-4-2-1-3-5-8/h1-7H
Inchi Key OLLKCCGWRITPOV-UHFFFAOYSA-N

Description

Synonym 4-Formyl-2-phenyl-1,3-thiazole, (4-Formyl-1,3-thiazol-2-yl)benzene
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 51-52
Product Link https://store.apolloscientific.co.uk/product/2-phenyl-13-thiazole-4-carboxaldehyde

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-phenyl-13-thiazole-4-carboxaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart