APO455828887 Apollo Scientific Ltd

4-(Hydroxymethyl)-2-phenyl-1,3-thiazole

  • MDL number MFCD02682017
  • Cas Number : 23780-13-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 67-70
Synonym (2-Phenyl-1,3-thiazol-4-yl)methanol
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=19490

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=19490
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart