APO455828895 Apollo Scientific Ltd

4-(1H-Imidazol-1-yl)benzyl alcohol

  • MDL number MFCD02682059
  • Cas Number : 86718-08-3
  • Molecular Weight : 174.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 110-112
Iupac Name [4-(1H-imidazol-1-yl)phenyl]methanol
Inchi Code 1S/C10H10N2O/c13-7-9-1-3-10(4-2-9)12-6-5-11-8-12/h1-6,8,13H,7H2
Inchi Key SRQXVPAXMMIUFH-UHFFFAOYSA-N

Description

Synonym [4-(1H-Imidazol-1-yl)phenyl]methanol
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 110-112
Product Link https://store.apolloscientific.co.uk/product/4-1h-imidazol-1-ylbenzyl-alcohol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-1h-imidazol-1-ylbenzyl-alcohol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart